Learn More
Invitrogen™ Medroxyprogesterone Acetate Polyclonal Antibody

Description
Working Concentration: 0.0938 μg/mL. Sensitivity: 10 ng/mL Medroxyprogesterone Acetate produces 99% inhibition in a competitive ELISA employing Medroxyprogesterone Acetate polyclonal antibody.
Specifications
Specifications
| Antigen | Medroxyprogesterone Acetate |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 20.88 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.6 |
| Gene Alias | C24H34O4; Depo-Provera; Metigestrona; Provera |
| Host Species | Sheep |
| Immunogen | Medroxyprogesterone acetate(3)-BTG. |
| Purification Method | purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.