Learn More
MEDTOX™ MEDTOXScan™ Positive and Negative QC Test Device
Catalog No. 22000405
Change view
Click to view available options
Intended Use:
Preprinted quality control devices used to check that the MEDTOXScan reader is functioning properly.
Catalog No. Intended Use
22-000-405 Preprinted quality control devices used to check that the MEDTOXScan reader is functioning properly.
1 options
Catalog No. 22-000-405 Supplier MEDTOX™ Supplier No. 833075
Add to Cart
Add to Cart
Detect errors associated with the MEDTOXScan™ Reader and a contaminated contact imaging sensor (CIS). MEDTOXScan™ Positive and Negative QC Test Devices verify that the CIS cleaning procedure using the MEDTOXScan™ Cleaning Cassette effectively removed any contamination.
  • Preprinted quality control devices used to check that the MEDTOXScan reader is functioning properly
  • For in vitro diagnostic use only
The QC Test Devices function as an optical performance system check for the MEDTOXScan Reader only, not the PROFILE™-V MEDTOXScan system, and are not intended to replace the need for the external controls.
The QC Test Devices have been designed to simulate the end points that are generated in the PROFILE-V MEDTOXScan test device when external positive and negative QC controls are run.
The QC Test Devices consist of artificial control lines and test lines (negative) or artificial control lines and no test lines (positive) printed on a membrane and placed in the PROFILE-V MEDTOXScan plastic housing.
The QC Test Devices are not intended to evaluate all components of the test system from specimen preparation through generation of results. They are intended to function as a troubleshooting device to determine that the reader optics are functioning correctly.
You should run the QC Test devices (1) if you suspect the MEDTOXScan Reader is not functioning properly, or (2) if you suspect the CIS is dirty, or (3) if the MEDTOXScan Reader has been dropped or damaged.
Specifications
Intended Use Preprinted quality control devices used to check that the MEDTOXScan reader is functioning properly.
Packaging Light blocking storage bag