Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEF2A Antibody (2F9H2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | MEF2A |
---|---|
Clone | 2F9H2 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
Applications | Western Blot, ELISA |
Description
MEF2A Monoclonal specifically detects MEF2A in Human samples. It is validated for Western Blot, ELISA.Specifications
MEF2A | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
PBS with 0.05% Sodium Azide | |
4205 | |
Purified recombinant fragment of human MEF2A (AA: 391-497) expressed in E. Coli. | |
Primary | |
54.8 kDa |
2F9H2 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
Monoclonal | |
Mouse | |
Human | |
ADCAD1, MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancerfactor 2A), MEF2, myocyte enhancer factor 2A, myocyte-specific enhancer factor 2A, RSRFC4, RSRFC9, Serum response factor-like protein 1 | |
MEF2A | |
IgG1 | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title