Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEF2A/MEF2C Recombinant Rabbit Monoclonal Antibody (10I9G9)
GREENER_CHOICE

Catalog No. PIMA542794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542794 Supplier Invitrogen™ Supplier No. MA542794
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: RD, Mouse brain. Immunogen sequence: NRKPDLRVLI PPGSKNTMPS VSEDVDLLLN QRINNSQSAQ SLATPVVSVA TPTLPGQGMG GYPSAISTTY GTEYSLSSAD LSSLSGFNTA SALHLGSVTG WQQQHLHNMP PSALSQLGAC TSTHLSQSSN LSLPSTQSLN IKSEPVSPPR DRTTTPSRYP QHTRHEAGRS PVDSLSSCSS SYDGSDREDH RNEFHSPIGL TRPSPDERES PSVKRMRLSE GWAT.

MEF2A is found in many muscle specific-genes mediating cellular functions in skeletal and cardiac muscle. MEF2C is a transcription activator which binds specifically to the MEF2 element present in the regulatory regions of many muscle-specific genes. Controls cardiac morphogenesis and myogenesis, and is also involved in vascular development. Plays an essential role in hippocampal-dependent learning and memory by suppressing the number of excitatory synapses and thus regulating basal and evoked synaptic transmission. Crucial for normal neuronal development, distribution, and electrical activity in the neocortex. Necessary for proper development of megakaryocytes and platelets and for bone marrow B lymphopoiesis. Required for B-cell survival and proliferation in response to BCR stimulation, efficient IgG1 antibody responses to T-cell-dependent antigens and for normal induction of germinal center B cells. May also be involved in neurogenesis and in the development of cortical architecture. Isoform 3 and isoform 4, which lack the repressor domain, are more active than isoform 1 and isoform 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEF2A/MEF2C
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 10I9G9
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MEF2A
Gene Accession No. Q02078, Q06413, Q60929, Q8CFN5
Gene Alias ADCAD1; C5DELq14.3; DEL5q14.3; mef2; NEDHSIL; RSRFC4; RSRFC9
Gene Symbols MEF2A, MEF2C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 250-473 of human MEF2A+MEF2C (Q06413).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17258, 17260, 4205, 4208
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.