Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEF2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568821 Supplier Invitrogen™ Supplier No. PA568821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

The proteins belonging to structural maintenance of chromosomes (SMC) family play critical roles in chromosome structural changes. SMC proteins are involved in the proper condensation and segregation of mitotic chromosomes. Four human SMC proteins that form two distinct heterodimeric complexes in the cell, the human chromosome-associated protein (hCAP)-C and hCAP-E protein complex (hCAP-C/hCAP-E), and the human SMC1 (hSMC1) and hSMC3 protein complex (hSMC1/hSMC3) have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEF2A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MEF2A
Gene Accession No. Q60929
Gene Alias A430079H05Rik; ADCAD1; C5DELq14.3; DEL5q14.3; MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancer factor 2A); MADS box transcription enhancer factor 2, polypeptide C; mef2; Mef2a; MEF2C; myocyte enhancer factor 2a; myocyte enhancer factor 2C; Myocyte-specific enhancer factor 2A; myocyte-specific enhancer factor 2C; RSRFC4; RSRFC9; Serum response factor-like protein 1
Gene Symbols MEF2A
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of mouse MEF2A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17258
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.