Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEF2B Antibody (PCRP-MEF2B-2F9) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314207
This item is not returnable.
View return policy
Description
MEF2B Monoclonal antibody specifically detects MEF2B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
MEF2B | |
Monoclonal | |
1 mg/mL | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug per 100-500 ug protein, Protein Array | |
LOC729991-MEF2B readthrough transcript, MEF2B, myocyte enhancer factor 2B | |
Recombinant full-length human MEF2B protein (Uniprot: Q02080) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array | |
PCRP-MEF2B-2F9 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
4207 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction