Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MEF2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552095
Description
Immunogen sequence: VLLKYTEYSE PHESRTNTDI LETLKRRGIG LDGPELEPDE GPEEPGEKFR RLAGEGGDPA LPRPRLYPAA PAMPSPDVVY GALPPPGCDP SGLGEALPAQ SRPSPFRPAA PKAGPPGLVH PLFSPSHLTS KTPPPLYLPT E Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 62%.
The myocyte enhancer factor-2 (MEF-2) family of transcription factors associated with co-repessors or co-activators to regulate development and function of T cells, neuronal cells and muscle cells. Four family members arise from alternatively spliced transcripts, termed MEF-2A, -2B, -2C and -2D. These members bind as homo- and heterodimers to the MEF-2 site in the promoter region of affected genes. Differential regulation in the expression of the four transcripts implies functional distinction for each during embryogenesis and development. The process of differentiation from mesodermal precursor cells to myoblasts has led to the discovery of a variety of tissue-specific factors that regulate muscle gene expression.
Specifications
MEF2B | |
Polyclonal | |
Unconjugated | |
MEF2B | |
2310045N01Rik; AI451606; BLOC-1-related complex subunit 8; Borcs8; BORCS8-MEF2B; BORCS8-MEF2B readthrough; LOC729991-MEF2B; LOC729991-MEF2B readthrough; MADS box transcription enhancer factor 2, polypeptide B (myocyte enhancer factor 2B); MEF2B; MEF2B neighbor gene protein homolog; Mef2bnb; MEF2BNB-MEF2B; MEF2BNB-MEF2B readthrough; myocyte enhancer factor 2B; myocyte-specific enhancer factor 2B; protein MEF2BNB; RIKEN cDNA 2310045N01 gene; RSRFR2; Serum response factor-like protein 2; XMEF2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100271849 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.30 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q02080 | |
MEF2B | |
Recombinant protein corresponding to Human MEF2B. Recombinant protein control fragment (Product #RP-102465). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction