Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEF2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575240
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575240 Supplier Invitrogen™ Supplier No. PA575240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is >95% (by SDS-PAGE).

The myocyte enhancer factor-2 (MEF-2) family of transcription factors associated with co-repessors or co-activators to regulate development and function of T cells, neuronal cells and muscle cells. Four family members arise from alternatively spliced transcripts, termed MEF-2A, -2B, -2C and -2D. These members bind as homo- and heterodimers to the MEF-2 site in the promoter region of affected genes. Differential regulation in the expression of the four transcripts implies functional distinction for each during embryogenesis and development. The process of differentiation from mesodermal precursor cells to myoblasts has led to the discovery of a variety of tissue-specific factors that regulate muscle gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEF2B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MEF2B
Gene Accession No. O55087, Q02080
Gene Alias 2310045N01Rik; AI451606; BLOC-1-related complex subunit 8; Borcs8; BORCS8-MEF2B; BORCS8-MEF2B readthrough; LOC729991-MEF2B; LOC729991-MEF2B readthrough; MADS box transcription enhancer factor 2, polypeptide B (myocyte enhancer factor 2B); MEF2B; MEF2B neighbor gene protein homolog; Mef2bnb; MEF2BNB-MEF2B; MEF2BNB-MEF2B readthrough; myocyte enhancer factor 2B; myocyte-specific enhancer factor 2B; protein MEF2BNB; RIKEN cDNA 2310045N01 gene; RSRFR2; Serum response factor-like protein 2; XMEF2
Gene Symbols MEF2B
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 80-120 of Human MEF2B.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100271849, 17259, 498607
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.