Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEF2BNB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561076
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561076 Supplier Invitrogen™ Supplier No. PA561076
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEEPEMQLKG KKVTDKFTES VYVLANEPSV ALYRLQEHVR RSLPELAQHK ADMQRWEEQS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MEF2BNB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BORCS8
Gene Accession No. Q96FH0
Gene Alias 2310045N01Rik; BLOC-1 related complex subunit 8; BLOC-1-related complex subunit 8; Borcs8; MEF2B neighbor; MEF2B neighbor gene protein; MEF2B neighbor gene protein homolog; MEF2BNB; protein MEF2BNB; RIKEN cDNA 2310045N01 gene
Gene Symbols BORCS8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MEF2BNB. Recombinant protein control fragment (Product #RP-100562).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729991
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.