Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEF2C Antibody (SR1855), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | MEF2C |
---|---|
Clone | SR1855 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
MEF2C Monoclonal antibody specifically detects MEF2C in Human, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
MEF2C | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
DEL5q14.3, myocyte enhancer factor 2C, myocyte-specific enhancer factor 2C, polypeptide C | |
A synthesized peptide derived from human MEF2C (Uniprot #: Q06413) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1855 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
4208 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title