Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEF2C Monoclonal Antibody (OTI1D1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502501S


Description
MEF2C Monoclonal antibody specifically detects MEF2C in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
MEF2C | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q06413 | |
Mouse | |
Affinity chromatography | |
RUO | |
4208 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1D1 | |
Unconjugated | |
MEF2C | |
5430401D19Rik; 9930028G15Rik; AV011172; C5DELq14.3; DEL5q14.3; hoo; hoover; id:ibd5026; MADS box transcription enhancer factor 2, polypeptide C; Mef2; mef2c; mef2c protein; mef2ca; Myocyte enhancer factor 2C; myocyte enhancer factor 2C MEF2C; myocyte enhancer factor 2ca; myocyte enhancer factor 2ca 4'-6'; myocyte enhancer factor 2ca delta gamma-like; myocyte enhancer factor 2ca variant 4; myocyte enhancer factor 2ca variant 5; myocyte enhancer factor 2ca variant 6; myocyte enhancer factor 2ca; myocyte-specific enhancer factor 2C; myocyte-specific enhancer factor 2C; RGD1563119; wu:fc05b06; zgc:64184; zgc:85726 | |
Full length human recombinant protein of human MEF2C produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction