Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEF2D Antibody (PCRP-MEF2D-3A4), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314120MFV500
Description
MEF2D Monoclonal antibody specifically detects MEF2D in Human samples. It is validated for Flow CytometrySpecifications
MEF2D | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
PCRP-MEF2D-3A4 | |
Flow Cytometry | |
DKFZp686I1536, MADS box transcription enhancer factor 2, polypeptide D (myocyte enhancerfactor 2D), MEF2D/DAZAP1 fusion, myocyte enhancer factor 2D, myocyte enhancer factor 2D/deleted in azoospermia associated protein 1 fusionprotein, myocyte-specific enhancer factor 2D | |
Recombinant full-length human MEF2D protein (Uniprot: Q14814) | |
0.1 mL | |
Phospho Specific | |
4209 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction