Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEF2D Monoclonal Antibody (OTI3D12)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526890

Catalog No. PIMA526890


Only null left
Add to Cart

Description

Description

MEF2D Monoclonal Antibody for Western Blot, IHC (P)

This gene is a member of the myocyte-specific enhancer factor 2 (MEF2) family of transcription factors. Members of this family are involved in control of muscle and neuronal cell differentiation and development, and are regulated by class II histone deacetylases. Fusions of the encoded protein with Deleted in Azoospermia-Associated Protein 1 (DAZAP1) due to a translocation have been found in an acute lymphoblastic leukemia cell line, suggesting a role in leukemogenesis. The encoded protein may also be involved in Parkinson disease and myotonic dystrophy. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MEF2D
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q14814
MEF2D
Human recombinant protein fragment corresponding to amino acids 1-282 of MEF2D produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI3D12
Unconjugated
MEF2D
C80750; DKFZp686I1536; MADS box transcription enhancer factor 2, polypeptide D; Mef2d; MEF2D/DAZAP1 fusion; myocyte enhancer factor 2D; myocyte-specific enhancer factor 2D
Mouse
Affinity Chromatography
RUO
4209
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.