Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEF2D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582282
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582282 Supplier Invitrogen™ Supplier No. PA582282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QPQPPQQQPP QPQQPQPQQP QQPQQPPQQQ SHLVPVSLSN LIPGSPLPHV GAALTVTTHP HISIKSEPVS PSRERSPAPP PPAVFPAARP EPGDGLSSPA GGSYETGDRD DGRGDFGPTL GLLRPAPEPE AEGSAVKRMR LDTWTL Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 95%.

This gene is a member of the myocyte-specific enhancer factor 2 (MEF2) family of transcription factors. Members of this family are involved in control of muscle and neuronal cell differentiation and development, and are regulated by class II histone deacetylases. Fusions of the encoded protein with Deleted in Azoospermia-Associated Protein 1 (DAZAP1) due to a translocation have been found in an acute lymphoblastic leukemia cell line, suggesting a role in leukemogenesis. The encoded protein may also be involved in Parkinson disease and myotonic dystrophy. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEF2D
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MEF2D
Gene Accession No. O89038, Q14814, Q63943
Gene Alias C80750; DKFZp686I1536; MADS box transcription enhancer factor 2, polypeptide D; Mef2d; MEF2D/DAZAP1 fusion; myocyte enhancer factor 2D; myocyte-specific enhancer factor 2D
Gene Symbols MEF2D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MEF2D. Recombinant protein control fragment (Product #RP-102532).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17261, 4209, 81518
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.