Learn More
MEF2D Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303521AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
This gene is a member of the myocyte-specific enhancer factor 2 (MEF2) family of transcription factors. Members of this family are involved in control of muscle and neuronal cell differentiation and development, and are regulated by class II histone deacetylases. Fusions of the encoded protein with Deleted in Azoospermia-Associated Protein 1 (DAZAP1) due to a translocation have been found in an acute lymphoblastic leukemia cell line, suggesting a role in leukemogenesis. The encoded protein may also be involved in Parkinson disease and myotonic dystrophy. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Oct 2012].Specifications
MEF2D | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q14814 | |
MEF2D | |
Between 425 and 475 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
MEF2D | |
MADS box transcription enhancer factor 2, polypeptide D (myocyte enhancer factor 2D), MEF2D/DAZAP1 fusion, myocyte enhancer factor 2D/deleted in azoospermia associated protein 1 fusion protein, myocyte-specific enhancer factor 2D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4209 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.