Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mefenamic acid, 98%

Catalog No. aaj6270514
Click to view available options
Quantity:
25 g
100 g

CAS: 61-68-7 | C15H15NO2 | 241.29 g/mol

Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.

Solubility
It is soluble in acetone, chloroform, dichloromethane, methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-68-7
Molecular Formula C15H15NO2
Molecular Weight (g/mol) 241.29
MDL Number MFCD00051721
InChI Key HYYBABOKPJLUIN-UHFFFAOYSA-N
Synonym mefenamic acid, ponstel, ponstan, 2-2,3-dimethylphenyl amino benzoic acid, mephenamic acid, parkemed, coslan, mefacit, ponalar, methenamic acid
PubChem CID 4044
ChEBI CHEBI:6717
IUPAC Name 2-(2,3-dimethylanilino)benzoic acid
SMILES CC1=C(C(=CC=C1)NC2=CC=CC=C2C(=O)O)C

Specifications

Color White
Quantity 25 g
Merck Index 14,5798
Solubility Information It is soluble in acetone,chloroform,dichloromethane,methanol. Insoluble in water.
Formula Weight 241.29
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Mefenamic acid
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 200-513-1
RTECSNumber CB4550000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.