Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEFV Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP280113AF594
Description
MEFV Polyclonal antibody specifically detects MEFV in Human samples. It is validated for Western BlotSpecifications
MEFV | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Epitope affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 594 | |
FMF, FMFmarenostrin, Marenostrin, Mediterranean fever, MEFpyrin, MGC126560, MGC126586, TRIM20 | |
Synthetic peptide corresponding to aa 2-29 (A2KTPSDHLLSTLEELVPYDFEKFKFKLQ29) of human MEFV. | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity, Signal Transduction | |
4210 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction