Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MEFV Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518410
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene encodes a protein, also known as pyrin or marenostrin, that is an important modulator of innate immunity. Mutations in this gene are associated with Mediterranean fever, a hereditary periodic fever syndrome.
Specifications
| MEFV | |
| Polyclonal | |
| Unconjugated | |
| MEFV | |
| FMF; HGNC:6998; Marenostrin; Mediterranean fever; MEF; MEFV; MGC126560; MGC126586; Pyrin; TRIM20 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 4210 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O15553 | |
| MEFV | |
| Synthetic peptide sequence (EHLKKLRKSGEEQ) corresponding to the internal amino acids of MEFV (aa 428-441). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction