Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583579 Supplier Invitrogen™ Supplier No. PA583579
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LATWNFQFLP QVNGHPDPFD EIILFSLPPA LDWDSVHVPG PHAMTIQELV DYVNARQKQG IYEEYEDIRR ENPVGTFHCS MSPGNL Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 97%.

MEG2 is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an N-terminal domain that shares a significant similarity with yeast SEC14, which is a protein that has phosphatidylinositol transfer activity and is required for protein secretion through the Golgi complex in yeast. This PTP was found to be activated by polyphosphoinositide, and is thought to be involved in signaling events regulating phagocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEG2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTPN9
Gene Accession No. P43378
Gene Alias Meg2; meg-2; protein tyrosine phosphatase, non-receptor type 9; protein-tyrosine phosphatase MEG2; Protein-tyrosine phosphatase, non-receptor type 9 (Protein-tyrosine phosphatase MEG2) (PTPase-MEG2); PTPase MEG2; PTPase-MEG2; PTPMEG2; PTPN9; Tyrosine-protein phosphatase non-receptor type 9
Gene Symbols PTPN9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTPN9. Recombinant protein control fragment (Product #RP-98128).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5780
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.