Learn More
MSA™ Mega Swivel Anchor Kit
Catalog No. 19166586 Shop All MSA Products
Catalog No. 19-166-586 Supplier MSA™ Supplier No. 10129084
MSA's Anchorage Connector Components for use in fall arrest and confined space entry applications.
  • Removable Concrete Anchors are designed for use as anchorage connectors in concrete applications
  • Anchorage Connector Extension features a 3,600 lb rated snaphook on one end and a 3 in. O-ring on the other
  • Bolt D-ring with Hole Anchorage Connector is a permanent anchor connector designed for coupling a single fall arrest system to an anchorage such as beams or girders
  • Chain Anchorage Connector is a temporary component for use near sharp edges or in high heat
  • D-Plate Anchorage Connector provides a permanent anchorage point
  • Large Hook with Strap is a temporary, overhead anchorage connector
  • ManSafe Freestanding Constant Force Post is tested and approved for use on concrete, single ply membrane, asphalt sanded, asphalt stone-chipped, and steel roof profiles
Specifications
Product Type 10K MEGA Swivel Anchorage Connector Kit, Zinc-plated for STEEL
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.