Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEGAComfort MegaSole™, Gel Enhanced Dual Layer Memory Foam Insole

Catalog No. 18813508
Encompass
Click to view available options
Size:
Men's: 10-11, Women's: 12-13
Men's: 12-13
Men's: 14-15
Men's: 6-7, Women's: 8-9
Men's: 8-9, Women's: 10-11
Women's: 5-7

Ergonomically designed by podiatrists, Dual layer 100% memory foam, Clinically proven field tested to reduce pain and fatigue, Max shock absorption comfort

  • Uses patented combination of high-density dual layer 100% memory foam
  • Clinically proven and field tested to reduce pain and fatigue while increasing balance and comfort
  • Provides vibration reduction, heel and arch cushioning, massaging and aeration
    • Holes help to regulate foot temperature in footwear for all-day comfort and improve balance reactions
  • Ideal for stationary, mobile and working outdoors
  • For use with casual and work footwear
  • Silver ion anti-microbial technology in top cloth offering anti-bacterial, anti- perspiration and anti-odor
  • Machine washable
  • MEGAGel™ Visco technology

Specifications

Color Black, Blue, Yellow
For Use With (Application) Stationary, mobile and working outdoors
For Use With (Equipment) Casual and work footwear
Material Memory Foam
Size Men's: 10-11, Women's: 12-13
Gender Unisex
Waterproof No
Product Type MEGASole Insole
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.