Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEGAComfort MegaSole™, Gel Enhanced Dual Layer Memory Foam Insole

Ergonomically designed by podiatrists, Dual layer 100% memory foam, Clinically proven field tested to reduce pain and fatigue, Max shock absorption comfort

$38.66 - $42.48

Specifications

Color Black, Blue, Yellow
For Use With (Application) Stationary, mobile and working outdoors
For Use With (Equipment) Casual and work footwear
Material Memory Foam
Gender Unisex
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-813-505
Encompass
View Documents
MEGAComfort
MSW567
Women's: 5-7
Pair for $38.66
Only null left
Add to Cart
 
18-813-506
Encompass
View Documents
MEGAComfort
MSM67/W89
Men's: 6-7, Women's: 8-9
Pair for $42.48
Only null left
Add to Cart
 
18-813-507
Encompass
View Documents
MEGAComfort
MSM89/W1011
Men's: 8-9, Women's: 10-11
Pair for $38.66
Only null left
Add to Cart
 
18-813-508
Encompass
View Documents
MEGAComfort
MSM1011/W1213
Men's: 10-11, Women's: 12-13
Pair for $38.66
Only null left
Add to Cart
 
18-813-510
Encompass
View Documents
MEGAComfort
MSM1415
Men's: 14-15
Pair for $38.66
Only null left
Add to Cart
 
18-813-509
View Documents
MEGAComfort
MSM1213
Men's: 12-13 N/A N/A N/A
Description

Description

  • Uses patented combination of high-density dual layer 100% memory foam
  • Clinically proven and field tested to reduce pain and fatigue while increasing balance and comfort
  • Provides vibration reduction, heel and arch cushioning, massaging and aeration
    • Holes help to regulate foot temperature in footwear for all-day comfort and improve balance reactions
  • Ideal for stationary, mobile and working outdoors
  • For use with casual and work footwear
  • Silver ion anti-microbial technology in top cloth offering anti-bacterial, anti- perspiration and anti-odor
  • Machine washable
  • MEGAGel™ Visco technology
Specifications

Specifications

Black, Blue, Yellow
Casual and work footwear
Unisex
MEGASole Insole
Stationary, mobile and working outdoors
Memory Foam
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.