Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Megestrol Acetate
GREENER_CHOICESDP

Catalog No. 628550 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-855-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 62-855-0 Supplier R&D Systems™ Supplier No. 6285/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Synthetic progesterone analog

Synthetic progesterone analog. Dose-dependently inhibits growth of HepG2 cells (IC50 = 260 μM) in vitro and in vivo and reduces tumor volume. High dosages improve weight gain in tumor-bearing nude mice. Orally bioavailable.

Chemical Identifiers

CAS 595-33-5
Molecular Formula C24H32O4

Specifications

Quantity 50 mg
Solubility Soluble to 50mM in DMSO and to 20mM in ethanol
Product Line Tocris
Formula Weight 384.51
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 384.516
Chemical Name or Material 17-(Acetyloxy)-6-methylpregna-4,6-diene-3,20-dione
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.