Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEGF10 Polyclonal Antibody, Biotin, Bioss
SDP

Catalog No. 502281709
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1709 Supplier Bioss Supplier No. BS24335RBIOTIN
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MEGF10 Polyclonal antibody specifically detects MEGF10 in Human samples. It is validated for Western Blot

Specifications

Antigen MEGF10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene MEGF10
Gene Accession No. Q96KG7
Gene Alias 3000002B06Rik; EMARDD; Gm331; KIAA1780; MEGF10; multiple EGF like domains 10; multiple EGF-like domains 10; multiple EGF-like domains protein 10; multiple EGF-like-domains 10; multiple epidermal growth factor-like domains protein 10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MEGF10
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84466
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.