Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEGF11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA540109
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA540109 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA540109 Supplier Invitrogen™ Supplier No. PA540109
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody may recoginize two isoforms: isoform 1 (110 kDa) and isoform 2 (101 kDa). Recommended positive controls: mouse eye tissue. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May regulate the mosaic spacing of specific neuron subtypes in the retina through homotypic retinal neuron repulsion. Mosaics provide a mechanism to distribute each cell type evenly across the retina, ensuring that all parts of the visual field have access to a full set of processing elements. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEGF11
Applications Western Blot
Classification Polyclonal
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% Thimerosal; pH 7
Gene MEGF11
Gene Accession No. A6BM72, Q80T91
Gene Alias 2410080H04Rik; D130061K05; KIAA1781; Megf11; multiple EGF like domains 11; multiple EGF-like domains protein 11; multiple EGF-like-domains 11; multiple epidermal growth factor-like domains protein 11; UNQ1949/PRO4432
Gene Symbols MEGF11
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human MEGF11. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214058, 84465
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.