Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MEGF6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568234
Description
Predicted to react with Mouse and Rat samples.
MEGF6 gene ontology annotations related to this gene include calcium ion binding.
Specifications
MEGF6 | |
Polyclonal | |
Unconjugated | |
MEGF6 | |
2600001P17Rik; Egfl3; EGF-like domain-containing protein 3; EGF-like protein 3; EGF-like-domain, multiple 3; Epidermal growth factor-like protein 3; KIAA0815; MEGF6; multiple EGF like domains 6; multiple EGF-like domain protein 3; multiple EGF-like domains protein 6; multiple EGF-like-domains 6; multiple epidermal growth factor-like domains 6; multiple epidermal growth factor-like domains protein 6 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1953, 230971, 65049 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O75095, O88281, Q80V70 | |
MEGF6 | |
Synthesized peptide derived from human MEGF6. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction