Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEGF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103934 Supplier Invitrogen™ Supplier No. PA5103934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MEGF6.

MEGF6 gene ontology annotations related to this gene include calcium ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEGF6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MEGF6
Gene Accession No. O75095, O88281, Q80V70
Gene Alias 2600001P17Rik; Egfl3; EGF-like domain-containing protein 3; EGF-like protein 3; EGF-like-domain, multiple 3; Epidermal growth factor-like protein 3; KIAA0815; MEGF6; multiple EGF like domains 6; multiple EGF-like domain protein 3; multiple EGF-like domains protein 6; multiple EGF-like-domains 6; multiple epidermal growth factor-like domains 6; multiple epidermal growth factor-like domains protein 6
Gene Symbols MEGF6
Host Species Rabbit
Immunogen A synthesized peptide derived from human MEGF6(Accession O75095), corresponding to amino acid residues P141-F191.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1953, 230971, 65049
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.