Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEGF8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | MEGF8 |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MEGF8 Polyclonal specifically detects MEGF8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MEGF8 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
1954 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: PCSPMPRSPEECRRLRTCSECLARHPRTLQPGDGEASTPRCKWCTNCPEGACIGRNGSCTSENDCRINQREVFWAGNCSEAACGAADCEQCTREGKCMWTRQFKRTGETRRILSVQPTYDWTCFSHSLLNVS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
C19orf49, chromosome 19 open reading frame 49, EGFL4MGC120684, EGF-like domain-containing protein 4, EGF-like protein 4, EGF-like-domain, multiple 4, Epidermal growth factor-like protein 4, FLJ22365, HBV pre-s2 binding protein 1, KIAA0817, MGC138147, multiple EGF-like domain protein 4, Multiple EGF-like domains protein 8, multiple EGF-like-domains 8, multiple epidermal growth factor-like domains 8, multiple epidermal growth factor-like domains protein 8, SBP1 | |
MEGF8 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title