Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meglumine, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18604284 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-284 100 g Poly Bottle
18-604-285 500 g Poly Bottle
18-604-286 2.5 kg Poly Bottle
18-604-287 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-284 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1122100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

M1122, 6284-40-8, C7H17NO5

Spectrum™ Chemical Meglumine, USP is used as a functional excipient where it acts as a counterion to enhance API stability and solubility. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6284-40-8
Molecular Formula C7H17NO5
Molecular Weight (g/mol) 195.22
InChI Key MBBZMMPHUWSWHV-UHFFFAOYNA-N
IUPAC Name 6-(methylamino)hexane-1,2,3,4,5-pentol
SMILES CNCC(O)C(O)C(O)C(O)CO

Specifications

CAS Max % 100%
Boiling Point 210°C (410°F)
Ignition Residue 0.1%
Loss on Drying 1%
Melting Range 128-132°C
Packaging Poly Bottle
Quantity 100 g
Specific Rotation -1°5.7° to -1°7.3°
Percent Purity 99 to 100.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Meglumine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.