Learn More
Invitrogen™ MEI1 Polyclonal Antibody

Description
Immunogen sequence: KADSPRTALL CSAWLLTASF SAQQHKGSLQ VHQTLSVEMD QVLKALSFPK KKAALLSAAI LCFLRTALRQ SFSSALVALV PSGAQPLPAT KDTVLAPLRM SQVRSLVIGL QNLLVQKDPL LSQACVGCLE ALLDYLDARS PDIALHVASQ PWNRFLLFTL LDAGENSFLR P; Positive Samples: Rat brain.
Specifications
Specifications
| Antigen | MEI1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.73 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | MEI1 |
| Gene Accession No. | Q5TIA1, Q9D4I2 |
| Gene Alias | 4932408F18Rik; MEI1; meiosis defective 1; meiosis defective protein 1; meiosis inhibitor 1; meiosis inhibitor protein 1; meiotic double-stranded break formation protein 1; SPATA38; spermatogenesis associated 38 |
| Gene Symbols | MEI1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.