Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEI4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112882
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112882 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112882 Supplier Invitrogen™ Supplier No. PA5112882
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MEI4 Polyclonal Antibody for Western Blot, ELISA

MEI4 (Meiotic Double-Stranded Break Formation Protein 4) is required for DNA double-strand break formation in unsynapsed regions during meiotic recombination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEI4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MEI4
Gene Accession No. A8MW99, Q8BRM6
Gene Alias 4930486G11Rik; MEI4; meiosis-specific 4 homolog; Meiosis-specific protein MEI4; meiosis-specific protein MEI4-like; meiosis-specific, MEI4 homolog (S. cerevisiae); meiotic double-stranded break formation protein 4; UPF0623 protein
Gene Symbols MEI4
Host Species Rabbit
Immunogen Recombinant Human Meiosis-specific protein MEI4-like protein (22-137AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101928601, 75033
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.