Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEIG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560769 Supplier Invitrogen™ Supplier No. PA560769
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASSDVKPTS VSHAKKWSEE IENLYRFQQA GYRDETEYRQ VKQVSMVDRW PETGYVKKLQ RRDNTFYYYN KQRECDDKEV HKVK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.

MEIG1, a murine gene first identified as a testis specific gene, is a chromosome/chromatin-binding protein initially expressed during meiosis but retained in the germ cell nucleus throughout later stages of spermatogenesis. MEIG1 is a highly conserved basal metazoan gene that is indispensable for mouse spermatogenesis. It is important for normal meiotic differentiation and absolutely crucial for terminal differentiation of spermatozoa. MEIG1 encodes two alternative transcripts, designated 2a2 and 11a2, both of which encode for a common ORF but differing in their 5' untranslated region (5'UTR) due to alternative promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEIG1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MEIG1
Gene Accession No. Q5JSS6
Gene Alias bA2K17.3; Meg1; Meig1; meiosis expressed gene 1; meiosis expressed gene 1 homolog; meiosis expressed gene 1 protein homolog; meiosis/spermiogenesis associated 1; Meiosis-expressed gene 1 protein; MLZ-278; Protein expressed in male leptotene and zygotene spermatocytes 278; SPATA39; spermatogenesis associated 39
Gene Symbols MEIG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MEIG1. Recombinant protein control fragment (Product #RP-97201).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644890
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.