Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEIKIN Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144240
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144240 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144240 Supplier Invitrogen™ Supplier No. PA5144240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MEIKIN Polyclonal Antibody for IHC (P), ELISA

Key regulator of kinetochore function during meiosis I: required both for mono-orientation of kinetochores on sister chromosomes and protection of centromeric cohesin from separase-mediated cleavage. Acts by facilitating kinetochore mono-orientation during meiosis I, when kinetochores on sister chromosomes face the same direction and are thus captured and pulled by spindle fibers from the same pole. Also required to prevent cleavage of cohesin at centromeres during meiosis I, possibly by acting as a regulator of the shugoshin-dependent protection pathway. Acts in collaboration with PLK1: required for PLK1 enrichment to kinetochores. Not required during meiosis II or mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEIKIN
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene MEIKIN
Gene Accession No. A0A087WXM9
Gene Alias MEIKINMeiosis-specific kinetochore protein
Gene Symbols MEIKIN
Host Species Rabbit
Immunogen Recombinant Human Meiosis-specific kinetochore protein (18-91AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728637
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.