Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEIOB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568422
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568422 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568422 Supplier Invitrogen™ Supplier No. PA568422
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MEIOB Polyclonal Antibody for IHC (P)

Single-stranded DNA-binding protein required for homologous recombination in meiosis I. Required for double strand breaks (DSBs) repair and crossover formation and promotion of faithful and complete synapsis. Not required for the initial loading of recombinases but required to maintain a proper number of RAD51 and DMC1 foci after the zygotene stage. May act by ensuring the stabilization of recombinases, which is required for successful homology search and meiotic recombination. Displays Single-stranded DNA 3'-5' exonuclease activity in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEIOB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MEIOB
Gene Accession No. Q8N635
Gene Alias 4930528F23Rik; C16orf73; C6H16orf73; gs129; Meiob; meiosis specific with OB domains; meiosis specific with OB-fold; meiosis-specific with OB domain-containing protein; MLZ-675; Protein expressed in male leptotene and zygotene spermatocytes 675
Gene Symbols MEIOB
Host Species Rabbit
Immunogen Synthetic protein corresponding to residues near the C terminal of human MEIOB.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254528
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.