Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Active Motif MEIS 1/2/3 Monoclonal Antibody (9.2.7)
Mouse Monoclonal Antibody
Specifications
Antigen | MEIS 1/2/3 |
---|---|
Clone | 9.2.7 |
Concentration | 1 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | Immunocytochemistry |
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
SPN-39796
|
10 μg | N/A | N/A | N/A | |||||
Description
MEIS 1/2/3 Monoclonal Antibody for ICC/IF
These gene encode homeobox proteins belonging to the TALE family of homeodomain-containing proteins. TALE homeobox proteins are highly conserved transcription regulators, and several members have been shown to be essential contributors to developmental programs. Multiple transcript variants encoding distinct isoforms have been described for this gene.Specifications
MEIS 1/2/3 | |
1 mg/mL | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
70mM tris with 0.07mM EDTA, 30% glycerol, 31mM glycine, 105mM NaCl and 0.035% sodium azide; pH 8 | |
O00470, O14770, P97367, P97368, Q60954, Q99687 | |
17268, 17536, 17537, 4211, 4212, 56917 | |
This MEIS antibody was raised against a recombinant protein corresponding to amino acids amino acids 50-390 mouse MEIS1. | |
Antibody |
9.2.7 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Monoclonal | |
Liquid | |
IgG1 | |
Human, Mouse | |
MEIS2 | |
A430109D20Rik; AI573393; C530044H18Rik; Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2; Evi8; Homeobox protein Meis1; homeobox protein Meis2; Homeobox protein Meis3; HsT18361; leukemogenic homolog protein; Meis homeobox 1; Meis homeobox 2; Meis homeobox 3; Meis homolog 2; MEIS1; Meis1, myeloid ecotropic viral integration site 1 homolog; Meis1, myeloid ecotropic viral integration site 1 homolog 2; Meis1, myeloid ecotropic viral integration site 1 homolog 3; Meis1-related gene 1; meis1-related protein 1; Meis1-related protein 2; MEIS2; Meis2 homeobox; MEIS3; Mrg1; Mrg2; Myeloid ecotropic viral integration site 1; myeloid ecotropic viral integration site-related gene 1; Stra10; TALE homeobox protein Meis2; WUGSC:H_NH0444B04.1 | |
MEIS1, MEIS2, MEIS3 | |
Primary | |
Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title