Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEIS 1/2/3 Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™
SDP

Catalog No. 501993782
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3782 10 μg
50-199-3781 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3782 Supplier Active Motif Supplier No. 91260
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

These gene encode homeobox proteins belonging to the TALE family of homeodomain-containing proteins. TALE homeobox proteins are highly conserved transcription regulators, and several members have been shown to be essential contributors to developmental programs. Multiple transcript variants encoding distinct isoforms have been described for this gene.

Specifications

Antigen MEIS 1/2/3
Applications Western Blot
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5
Gene MEIS1
Gene Accession No. O00470, O14770, P97367, P97368, Q60954, Q99687
Gene Symbols MEIS1, MEIS2, MEIS3
Host Species Mouse
Immunogen This antibody was raised against recombinant protein comprising amino acids 59-390 of mouse MEIS1.
Purification Method Protein A
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17268, 17536, 17537, 4211, 4212, 56917
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.