Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEIS1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$207.00 - $426.00

Specifications

Antigen MEIS1
Concentration 0.5 mg/mL
Dilution Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224597SS
SDP
View Documents
Novus Biologicals
NBP224597SS
0.025 mg
Each for $207.00
Only null left
Add to Cart
 
NBP224597
SDP
View Documents
Novus Biologicals
NBP224597
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

MEIS1 Polyclonal specifically detects MEIS1 in Human, Mouse, Rat, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

MEIS1
Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Polyclonal
Purified
RUO
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
homeobox protein Meis1, leukemogenic homolog protein, Meis homeobox 1, MGC43380, myeloid ecotropic viral integration site 1 homolog, myeloid ecotropic viral integration site 1 homolog (mouse)
MEIS1
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, rat, mouse, orangutan, bovine and chicken, 88% homologous in xenopus, and 73% homologous in zebrafish and pufferfish.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat, Chicken, Primate
O00470
4211
A portion of amino acids 170-220 of MEIS1 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.