Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEIS1 Monoclonal Antibody (OTI3G8)
GREENER_CHOICE

Catalog No. PIMA527190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527190 Supplier Invitrogen™ Supplier No. MA527190
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MEIS1 Monoclonal Antibody for Western Blot, IHC (P)

Homeobox genes, of which the most well-characterized category is represented by the HOX genes, play a crucial role in normal development. In addition, several homeoproteins are involved in neoplasia. This gene encodes a homeobox protein belonging to the TALE family of homeodomain-containing proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEIS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3G8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MEIS1
Gene Accession No. O00470, Q60954
Gene Alias C530044H18Rik; cb224; etID37540.13; Evi8; Homeobox protein Meis1; homeobox protein Meis1b; id:ibd5078; leukemogenic homolog protein; Meis homeobox 1; Meis homeobox 1 b; Meis homeobox 1 S homeolog; meis1; Meis1, myeloid ecotropic viral integration site 1 homolog; meis1.1; meis1.S; MEIS1A; meis1b; MGC43380; myeloid ecotropic viral insertion site-1a protein; myeloid ecotropic viral integration 1; myeloid ecotropic viral integration site 1; transcription factor Meis1.1; wu:fb38b03; wu:fc02e11; wu:fd12a02; WUGSC:H_NH0444B04.1; XMeis1; XMeis1-1 protein; XMeis1-2 protein; XMeis1-4 protein
Gene Symbols MEIS1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 176-390 of MEIS1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17268, 4211, 686117
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.