Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEIS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112605
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112605 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112605 Supplier Invitrogen™ Supplier No. PA5112605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MEIS1 Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA

Homeobox genes, of which the most well-characterized category is represented by the HOX genes, play a crucial role in normal development. In addition, several homeoproteins are involved in neoplasia. This gene encodes a homeobox protein belonging to the TALE family of homeodomain-containing proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEIS1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MEIS1
Gene Accession No. O00470, Q60954
Gene Alias C530044H18Rik; cb224; etID37540.13; Evi8; Homeobox protein Meis1; homeobox protein Meis1b; id:ibd5078; leukemogenic homolog protein; Meis homeobox 1; Meis homeobox 1 b; Meis homeobox 1 S homeolog; meis1; Meis1, myeloid ecotropic viral integration site 1 homolog; meis1.1; meis1.S; MEIS1A; meis1b; MGC43380; myeloid ecotropic viral insertion site-1a protein; myeloid ecotropic viral integration 1; myeloid ecotropic viral integration site 1; transcription factor Meis1.1; wu:fb38b03; wu:fc02e11; wu:fd12a02; WUGSC:H_NH0444B04.1; XMeis1; XMeis1-1 protein; XMeis1-2 protein; XMeis1-4 protein
Gene Symbols MEIS1
Host Species Rabbit
Immunogen Recombinant Human Homeobox protein Meis1 protein (1-90AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17268, 4211
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.