Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEIS2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298816AF350
Description
MEIS2 Polyclonal specifically detects MEIS2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MEIS2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
Alexa Fluor 350 | |
Meis (mouse) homolog 2, Meis homeobox 2, Meis1, myeloid ecotropic viral integration site 1 homolog 2 (mouse), Meis1-related gene 1, MGC2820, myeloid ecotropic viral integration site 1 homolog 2, TALE homeobox protein Meis2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human MEIS2. | |
0.1 mL | |
Vision | |
4212 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction