Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEIS2 Antibody (PCRP-MEIS2-2B3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320520100UG
Description
MEIS2 Monoclonal antibody specifically detects MEIS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
MEIS2 | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate, Immunohistochemistry-Paraffin 1-2 μg/mL | |
Meis (mouse) homolog 2, Meis homeobox 2, Meis1, myeloid ecotropic viral integration site 1 homolog 2 (mouse), Meis1-related gene 1, MGC2820, myeloid ecotropic viral integration site 1 homolog 2, TALE homeobox protein Meis2 | |
Recombinant fragment of human MEIS2 | |
100 μg | |
Vision | |
4212 | |
Store at 4&drg;C. Do not freeze. | |
IgG2a κ |
Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
PCRP-MEIS2-2B3 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction