Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MEIS2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519311
Description
This antibody is predicted to react with bovine, canine and human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a homeobox protein belonging to the TALE family of homeodomain-containing proteins. TALE homeobox proteins are highly conserved transcription regulators, and several members have been shown to be essential contributors to developmental programs. Multiple transcript variants encoding distinct isoforms have been described for this gene.
Specifications
| MEIS2 | |
| Polyclonal | |
| Unconjugated | |
| MEIS2 | |
| A430109D20Rik; Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2; homeobox protein Meis2; HsT18361; Meis homeobox 2; Meis homolog 2; Meis1, myeloid ecotropic viral integration site 1 homolog 2; Meis1-related gene 1; meis1-related protein 1; MEIS2; Meis2 homeobox; MGC2820; MRG1; myeloid ecotropic viral integration site-related gene 1; OTTHUMP00000160209; Stra10; TALE homeobox protein Meis2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 17536, 311311, 4212 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O14770, P97367 | |
| MEIS2 | |
| Synthetic peptide sequence (QSNRAGFLLDPSVSQ) corresponding to the internal amino acids of MEIS2 (aa 341-355). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction