Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MEIS2 Recombinant Rabbit Monoclonal Antibody (2A6I3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538081
Description
Positive Samples: SKOV3, U-937, U-87MG, Mouse lung, Mouse kidney Immunogen sequence: MAQRYDELPH YGGMDGVGVP ASMYGDPHAP RPIPPVHHLN HGPPLHATQH YGAHAPHPNV MPASMGSAVN DALKRDKDAI YGHPLFPLLA LVFEKCELAT.
This gene encodes a homeobox protein belonging to the TALE family of homeodomain-containing proteins. TALE homeobox proteins are highly conserved transcription regulators, and several members have been shown to be essential contributors to developmental programs. Multiple transcript variants encoding distinct isoforms have been described for this gene.
Specifications
MEIS2 | |
Recombinant Monoclonal | |
0.4 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14770, P97367 | |
MEIS2 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MEIS2 (O14770). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
2A6I3 | |
Unconjugated | |
MEIS2 | |
A430109D20Rik; Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2; homeobox protein Meis2; HsT18361; Meis homeobox 2; Meis homolog 2; Meis1, myeloid ecotropic viral integration site 1 homolog 2; Meis1-related gene 1; meis1-related protein 1; MEIS2; Meis2 homeobox; MGC2820; MRG1; myeloid ecotropic viral integration site-related gene 1; OTTHUMP00000160209; Stra10; TALE homeobox protein Meis2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
17536, 4212 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction