Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEIS3 Monoclonal Antibody (OTI3C9)
GREENER_CHOICE

Catalog No. PIMA526414
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526414 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526414 Supplier Invitrogen™ Supplier No. MA526414
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MEIS3 Monoclonal Antibody for Flow, IHC (P), Western Blot

Transcriptional regulator which directly modulates PDPK1 expression, thus promoting survival of pancreatic beta-cells. Also regulates expression of NDFIP1, BNIP3, and CCNG1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEIS3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MEIS3
Gene Accession No. Q99687
Gene Alias AI573393; Homeobox protein Meis3; Meis homeobox 3; Meis homeobox 3 pseudogene 1; Meis1, myeloid ecotropic viral integration site 1 homolog 3; meis1-related protein 2; Meis3; MEIS3P1; Mrg2; myeloid ecotropic viral integration site-related gene 2
Gene Symbols MEIS3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-261 of MEIS3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56917
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.