Learn More
Integra™ Miltex™ MeisterHand™ Metzenbaum (Lahey) Scissors
Standard pattern
Supplier: Integra™ Miltex™ MH5180
Description
- Stainless steel
- Non-glare matte finish
- Blunt points
- Tungsten carbide cutting edge in straight, with gold ring handles
- Length: 5.5 in. (14cm)
Order Info
Cold or chemical sterilization not recommended for Tungsten carbide scissors.
Specifications
Curved | |
14.3 cm | |
Blunt/Blunt |
5.625 in. | |
Surgical, Metzenbaum |