Learn More
R&D Systems™ MEK InhibitorTocris™et™
SDP
Catalog No. 22431R Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Set
Catalog No. Quantity
22-431-R 1 Set
1 options
This item is not returnable. View return policy
Catalog No. 22-431-R Supplier R&D Systems™ Supplier No. 2243/1 SET
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Selection of 3 MEK inhibitors (Cat. Nos. 1213, 1969 and 1144)
Contains the listed products which can be used to selectively inhibit MEK signalling. The products are supplied either as a solid (U0126) or pre-dissolved in DMSO at a stock concentration of 10 mM in 0.5 ml.
Specifications
Content And Storage -20°C
Quantity 1 Set
Product Type MEK Inhibitor Tocriset™
Product Line Tocris
Inhibitors MEK Inhibitor Tocriset™