Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK1/2 Antibody (SR1751), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32163250UL
Description
MEK1/2 Monoclonal antibody specifically detects MEK1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
MEK1/2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR1751 | |
Western Blot 1:5000-1:10000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:50-1:100 | |
CFC3, CFC4, dual specificity mitogen-activated protein kinase kinase 1, dual specificity mitogen-activated protein kinase kinase 2, ERK activator kinase 1, ERK activator kinase 2, MAP kinase kinase 2, MAPK/ERK kinase 1, MAPK/ERK kinase 2, MAPKK 1, MAPKK1, MAPKK2, MEK 1, MEK1, MEK2, MEL, mitogen-activated protein kinase kinase 2, p45, MKK1, MKK2, PRKMK1, PRKMK2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1) | |
A synthesized peptide derived from human MEK1/2 (Uniprot #: Q02750, P36507) | |
50 μg | |
Phospho Specific | |
5604 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction