Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK1/2 Antibody (SR1751), Novus Biologicals™
SDP

Catalog No. p-200006542 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258888 100 μg
NB260376 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258888 Supplier Novus Biologicals Supplier No. NBP321632100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MEK1/2 Monoclonal antibody specifically detects MEK1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation

Specifications

Antigen MEK1/2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SR1751
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:10000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:50-1:100
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias CFC3, CFC4, dual specificity mitogen-activated protein kinase kinase 1, dual specificity mitogen-activated protein kinase kinase 2, ERK activator kinase 1, ERK activator kinase 2, MAP kinase kinase 2, MAPK/ERK kinase 1, MAPK/ERK kinase 2, MAPKK 1, MAPKK1, MAPKK2, MEK 1, MEK1, MEK2, MEL, mitogen-activated protein kinase kinase 2, p45, MKK1, MKK2, PRKMK1, PRKMK2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1)
Host Species Rabbit
Immunogen A synthesized peptide derived from human MEK1/2 (Uniprot #: Q02750, P36507)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5604
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.