Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK1 Antibody (OTI1F5), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272643IR
Description
MEK1 Monoclonal specifically detects MEK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MEK1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAP2K1 | |
Full-length protein expressed in 293T cell transfected with human MEK1 expression vector. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1F5 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
dual specificity mitogen-activated protein kinase kinase 1, ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK1, MEK1MAPKK 1, mitogen-activated protein kinase kinase 1, MKK1MEK 1, PRKMK1EC 2.7.12.2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1) | |
Mouse | |
Affinity Purified | |
Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Phosphatase, Signal Transduction, Tyrosine Kinases | |
5604.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction