Learn More
MEK1 C-Term Monoclonal Antibody (13B6.G12), Rockland™

Mouse Monoclonal Antibody
$174.73 - $651.91
Specifications
Antigen | MEK1 C-Term |
---|---|
Clone | 13B6.G12 |
Concentration | 1 mg/mL |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0613
![]() ![]() |
Rockland Immunochemicals
200301GQ0 |
100 μg |
Each for $651.91
|
|
|||||
50-199-0614
![]() ![]() |
Rockland Immunochemicals
200301GQ0S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. This product is stable at 4°C as an undiluted liquid. For extended storage, aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Dilute only prior to immediate use. Reacts specifically with human MEK1. Cross reactivity is expected to occur with human, mouse, and rat based on sequence identity of the peptide immunogen. This antibody does not react with the MEK2 isoform.
MEK1/mitogen activated protein kinase/ERK kinase 1/microtubule-associated protein 2 kinase is a dual specificity kinase and catalyzes both a threonine and a tyrosine residue on MAP kinases ERK1 and ERK2. Mek1 is activated by phosphorylation of serine 218 and 222 residues by Raf1. It is known to be involved in the signaling during stress activate response, apoptosis and proliferative induction by cytokines. Mutations in the gene can lead to cardiofaciocutaneous syndrome.Specifications
MEK1 C-Term | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
CFC3; dual specificity mitogen activated protein kinase kinase 1; Dual specificity mitogen-activated protein kinase kinase 1; EC 2.7.12.2; ERK activator kinase 1; kinase MEK1; MAP kinase kinase 1; MAP kinase kinase or Erk Kinase, Dual specificity mitogen-activated protein kinase kinase, involved in ras mediated vulval induction, LEThal LET-537 (42.8 kD) (mek-2); MAP kinase/Erk kinase 1; MAP2K1; map2k1.L; MAPK/ERK kinase 1; MAPKK 1; MAPKK1; MEK 1; Mek1; MEK2; mek-2; MEKK1; mitogen activated protein kinase kinase 1; mitogen-activated protein kinase kinase 1; mitogen-activated protein kinase kinase 1 L homeolog; MKK (Thr292); MKK (Thr386); MKK1; MKK2; MP2K1; Prkmk1; PRKMK2; protein kinase, mitogen activated, kinase 1, p45; protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1); XELAEV_18018447mg | |
Map2k1 | |
IgG1 κ | |
-20°C, Avoid Freeze/Thaw Cycles | |
Map2k1 |
13B6.G12 | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P31938, Q01986, Q02750 | |
170851, 26395, 5604 | |
Anti-MEK1 Monoclonal Antibody was produced in mice by repeated immunizations with synthetic peptide corresponding to amino acid residues near the C-terminus conjugated to KLH. | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.